Learn More
Sino Biological G-CSFR / CD114 / CSF3R Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10218MM03200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human G-CSFR / CD114 (rh G-CSFR / CD114; Catalog#10218-HCCH; NP_000751.1; Met1-Pro621). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3303