Learn More
Sino Biological Human Leptin Protein DFS Item
Manufacturer:  Sino Biological 10221HNAE2
A DNA sequence encoding the mature form of human Leptin (NP_000221.1) (Val 22-Cys 167) was expressed, with an additional Met.
Catalog No. 50-161-4373