Learn More
Sino Biological Leptin / LEP Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10221RP02200
Produced in rabbits immunized with purified, recombinant Human Leptin (rh Leptin; Catalog#10221-HNAE; NP_000221.1; Val 22-Cys 167). Leptin specific IgG was purified by human Leptin affinity chromatography.
Catalog No. 50-172-3349