Learn More
Sino Biological Human LCN2 / NGAL Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10222H08H100
A DNA sequence encoding the human Lipocalin 2 precursor (NP_005555.2) (Met 1-Gly 198) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4376