Learn More
Sino Biological NAPSA/Napsin A Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102221T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NAPSA/Napsin A, and purified by antigen affinity chromatography.
Catalog No. 50-172-3353