Learn More
Sino Biological Human DCR3 / TNFRSF6B Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10224H02B200
A DNA sequence encoding the human DCR3 (O95407)(Met1-His300) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4379