Learn More
Sino Biological PLAUR / CD87 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102261T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Rat PLAUR / CD87, and purified by antigen affinity chromatography.
Catalog No. 50-172-3395