Learn More
Sino Biological Human ALK-2 / ACVR1 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10227H03H100
A DNA sequence encoding the extracellular domain (Met 1-Val 124) of human ALK2 (NP_001104537.1) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4380