Learn More
Sino Biological ALK-2 / ACVR1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10227T24200
Produced in rabbits immunized with purified, recombinant Human ALK-2 / ACVR1 (rh ALK-2 / ACVR1; Catalog#10227-H08B; Q04771; Met1-Val124). ALK-2 / ACVR1 specific IgG was purified by Human ALK-2 / ACVR1 affinity chromatography.
Catalog No. 50-172-3409