Learn More
Sino Biological DROSHA Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102288T3250
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DROSHA, and purified by antigen affinity chromatography.
Catalog No. 50-172-3419