Learn More
Sino Biological BTLA / CD272 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102297T3250
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse BTLA / CD272, and purified by antigen affinity chromatography.
Catalog No. 50-172-3431