Learn More
Sino Biological NPY / Pro-neuropeptide Y Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102298T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human NPY / Pro-neuropeptide Y, and purified by antigen affinity chromatography.
Catalog No. 50-172-3434