Learn More
Sino Biological GOLGA2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 102306T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human GOLGA2, and purified by antigen affinity chromatography.
Catalog No. 50-172-3457