Learn More
Sino Biological Human EphB4 / HTK Protein (His Tag) 100ug
Supplier:  Sino Biological 10235H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ala 539) of human EphB4 (NP_004435.3) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4398