Learn More
Sino Biological Human EphB4 / HTK Protein DFS Item
Manufacturer:  Sino Biological 10235HCCH200
The mature form of human EphB4 (NP_004435.3) extracellular domain (Met 1-Ala 539) with quinary-aa peptide (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-4404