Learn More
Sino Biological Human Fas Ligand / FASLG / CD95L Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10244H07Y20
A DNA sequence encoding human FASLG (NP_000630.1) (Pro134-Leu281) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4424