Learn More
Sino Biological Human Lefty2 / Lefty A Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10248H01H20
A DNA sequence encoding the mature form of human LeftyA (NP_003231.2) (Phe 78-Pro 366) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4431