Learn More
Sino Biological Human CLEC4N / CLEC6A / Dectin-2 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10250H01H50
A DNA sequence encoding the human DECTIN2 (NP_001007034) (Thr 42-Leu209) was expressed, fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4435