Learn More
Sino Biological Human IL11RA / IL11Rα Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10252H08H50
A DNA sequence encoding the extracellular domain of human IL11Rα (NP_004503.1) (Met 1-Val 363) was expressed, with a fused polyhistidine tag at the C-terminus.
Catalog No. 50-161-4437