Learn More
Sino Biological Human CD64 / FCGR1A Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10256H08H50
A DNA sequence encoding the extracellular domain of human CD64A (NP_000557.1) (Met 1-Pro 288) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4445