Learn More
Sino Biological Human CD32b / FCGR2B Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10259H08C50
A DNA sequence encoding (Ala46-Pro217) of human CD32b (P31994-1) was expressed,with a C-terminal polyhistidine tag.
Catalog No. 50-161-4457