Learn More
Sino Biological Human CD32b / FCGR2B Protein (His & AVI Tag) DFS Item
Manufacturer:  Sino Biological 10259H27H100
A DNA sequence encoding the extracellular domain (Ala 46-Ile 224) of mature human CD32b (NP_001002274.1) protein was fused with a signal peptide at the N-terminus and a c-terminal polyhistidine tagged AVI tag at the C-terminus.
Catalog No. 50-161-4460