Learn More
Sino Biological Human FGF9 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10262H01H5
A DNA sequence encoding the mature form of human fibroblast growth factor 9 (NP_002001.1) (Leu 4-Ser 208) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4468