Learn More
Sino Biological Human MMP12 / MMP-12 / HME Protein (catalytic domain) DFS Item
Manufacturer:  Sino Biological 10266HNAE100
A DNA sequence encoding human MMP12 (NP_002417.2) (Gly106-Asn268) was expressed with a N-terminal Met.
Catalog No. 50-161-4472