Learn More
Sino Biological Human Noggin / NOG Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10267H08H20
A DNA sequence encoding the human Noggin precursor (NP_005441.1) (Met 1-Cys 232) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4478