Learn More
Sino Biological Human FKBP12 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10268H08E200
A DNA sequence encoding the human FKBP12 (NP_463460) (Met 1-Glu 108) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4484