Learn More
Sino Biological Human Osteoprotegerin / TNFRSF11B Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10271H08H50
A DNA sequence encoding the human TNFRSF11B (NP_002537.3) (Met 1-Leu 401) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4497