Learn More
Sino Biological Human PDGF-C Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10273H01H5
A DNA sequence encoding the mature form of human PDGF-C (NP_057289.1) (Val 235-Gly 345) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4500