Learn More
Sino Biological Human PLGF Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10274H05H50
A DNA sequence encoding the human PGF (NP_002623.2) (Met1-Arg170) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-4506