Learn More
Sino Biological Human Galectin-3 / LGALS3 Protein, Low Endotoxin DFS Item
Manufacturer:  Sino Biological 10289HNAEE10
A DNA sequence encoding the human Galectin3 (P17931) (Met 1-Ile 250) was expressed and purified.
Catalog No. 50-161-4531