Learn More
Sino Biological Human B7-DC / PD-L2 / CD273 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10292H08H200
A DNA sequence encoding the extracellular domain (Met 1-Pro 219) of human PD-L2 (NP_079515.2) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4543