Learn More
Sino Biological Human SerpinE1 / PAI-1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10296H08H10
A DNA sequence encoding the human SerpinE1 precursor (NP_000593.1) (Met 1-Pro 402) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4551