Learn More
Sino Biological Human MERTK / Mer Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10298H03H200
A DNA sequence encoding the extracellular domain (Met 1-Ala 499) of human Mer precursor (NP_006334.2) was fused with the polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4556