Learn More
Sino Biological Human MERTK / Mer Protein GST Tag DFS Item
Manufacturer:  Sino Biological 10298H20B150
A DNA sequence encoding the human MERTK (Q12866) protein kinase domain (Glu 578-Tyr 872) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-4558