Learn More
Sino Biological Human MERTK / Mer Protein DFS Item
Manufacturer:  Sino Biological 10298HCCH20
The mature form of human Mer (NP_006334.2) extracellular domain (Met 1-Ala 499) with five amino acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-4560