Learn More
Sino Biological Human Periostin / POSTN Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10299H08H50
A DNA sequence encoding the human POSTN isoform 3 (NP_001129407.1) corresponding to amino acid (Met 1-Gln 781) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4564