Learn More
Sino Biological Human PTP1B / PTPN1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10304H07E100
A DNA sequence encoding the human PTPN1 (NP_002818.1) (Glu 2-Asn 321) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-4572