Learn More
Sino Biological Human Leptin Receptor / LEPR / CD295 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10322H08H200
A DNA sequence encoding the extracellular domain (Met 1-Asp 839) of human leptin receptor (NP_002294.2) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-4591