Learn More
Sino Biological Human HAI-2 / SPINT2 Protein (ECD, Fc Tag) DFS Item
Manufacturer:  Sino Biological 10324H02H10
A DNA sequence encoding the human SPINT2 (NP_066925.1) (Met1-Lys197) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4594