Learn More
Sino Biological Human HMGB1 / HMG1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10326H07E20
A DNA sequence encoding the mature form of human HMGB1 (P09429) (Lys57-Asp158) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4603