Learn More
Sino Biological Human HMGB1 / HMG1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10326H08H50
A DNA sequence encoding the human HMGB1 protein (NP_002119.1) (Met 1-Glu 215) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-161-4606