Learn More
Sino Biological Human HVEM / TNFRSF14 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10334H03H200
A DNA sequence encoding the extracellular domain (Met 1-Val 202) of human HVEM (NP_003811.2) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4629