Learn More
Sino Biological Human E-Selectin / CD62e / SELE Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10335H03H100
A DNA sequence encoding the extracellular domain (Met 1-Pro 556) of human CD62E (NP_000441.2) precursor was expressed with the fused C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4633