Learn More
Sino Biological Human IFNα4 / IFNa4 / Interferon alpha-4 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10336H01H50
A DNA sequence encoding the mature form of human IFN-α4a (NP_066546.1) (Cys 23-Asp 189) was expressed with the N-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4639