Learn More
Sino Biological Human IFNGR1 / CD119 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10338H03H100
A DNA sequence encoding the extracellular domain (Met 1-Gly 245) of human IFN-γ receptor 1 pre-protein (NP_000407.1) was fused with C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4647