Learn More
Sino Biological Human Interferon alpha 7 / IFNA7 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10341H01H100
A DNA sequence encoding the human IFNA7 (P01567) (Cys 24-Asp 189) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4657