Learn More
Sino Biological Human Granzyme B / GZMB Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10345H08H10
A DNA sequence encoding the proform of human Granzyme B (NP_004122.1) (Met 1-Tyr 247) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4675