Learn More
Sino Biological Human ICAM-1 / CD54 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10346H03H100
A DNA sequence encoding the human ICAM1 (NP_000192.2) extracellular domain (Met 1-Glu 480) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4677