Learn More
Sino Biological Human ICAM-1 / CD54 Protein DFS Item
Manufacturer:  Sino Biological 10346HCCH100
A DNA sequence encoding the human ICAM1 (NP_000192.2) extracellular domain (Met 1-Glu 480) with five amino acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-4681