Learn More
Sino Biological Human Interferon alpha-B / IFNA8 Protein DFS Item
Manufacturer:  Sino Biological 10347HNCH50
A DNA sequence encoding the human IFNA8 (NP_002161.2)(Cys24-Glu189) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-161-4686